RIE156321122 Rieke Metals, Inc.

Hexyl 5-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylate

  • MDL number MFCD07699080
  • Cas Number : 898772-20-8
  • Molecular Weight : 284.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-02E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-02E
Availability details for
Quantity
  • Availability
  • Add to Cart