RIE156321124 Rieke Metals, Inc.

2-Trifluoroacetyl-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD08064093
  • Cas Number : 898772-29-7
  • Molecular Weight : 252.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-03
Availability details for
Quantity
  • Availability
  • Add to Cart