RIE156321125 Rieke Metals, Inc.

Ethyl 5-(1,3-Dioxolan-2-yl)-2-thenoylformate

  • MDL number MFCD07699084
  • Cas Number : 898772-32-2
  • Molecular Weight : 256.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-04
Availability details for
Quantity
  • Availability
  • Add to Cart