RIE156321129 Rieke Metals, Inc.

4-Chlorobutyl 5-(1,3-dioxolan-2-yl)-2-thienyl ketone

  • MDL number MFCD07699089
  • Cas Number : 898772-47-9
  • Molecular Weight : 274.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-05D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-05D
Availability details for
Quantity
  • Availability
  • Add to Cart