RIE156321131 Rieke Metals, Inc.

Cyclobutyl 5-(1,3-dioxolan-2-yl)-2-thienyl ketone

  • MDL number MFCD07699091
  • Cas Number : 898772-52-6
  • Molecular Weight : 238.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclobutyl[5-(1,3-dioxolan-2-yl)-2-thienyl]methanone
Inchi Code 1S/C12H14O3S/c13-11(8-2-1-3-8)9-4-5-10(16-9)12-14-6-7-15-12/h4-5,8,12H,1-3,6-7H2
Inchi Key MWOQIFQQAAFKLB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5567-06c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-06c
Availability details for
Quantity
  • Availability
  • Add to Cart