RIE156321136 Rieke Metals, Inc.

5-(1,3-dioxolan-2-yl)-2-thienyl 4-oxopentyl ketone

  • MDL number MFCD07699098
  • Cas Number : 898772-66-2
  • Molecular Weight : 268.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-07C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-07C
Availability details for
Quantity
  • Availability
  • Add to Cart