RIE156321137 Rieke Metals, Inc.

5-(1,3-dioxolan-2-yl)-2-thienyl 5-oxohexyl ketone

  • MDL number MFCD07699099
  • Cas Number : 898772-68-4
  • Molecular Weight : 282.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-07D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-07D
Availability details for
Quantity
  • Availability
  • Add to Cart