RIE156321138 Rieke Metals, Inc.

Cyclohexylmethyl 5-(1,3-dioxolan-2-yl)-2-thienyl ketone

  • MDL number MFCD07699101
  • Cas Number : 898772-72-0
  • Molecular Weight : 280.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-08A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-08A
Availability details for
Quantity
  • Availability
  • Add to Cart