RIE156321139 Rieke Metals, Inc.

(2-Cyclohexyl)ethyl 5-(1,3-dioxolan-2-yl)-2-thienyl ketone

  • MDL number MFCD07699102
  • Cas Number : 898772-74-2
  • Molecular Weight : 294.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-08B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-08B
Availability details for
Quantity
  • Availability
  • Add to Cart