RIE156321144 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl 2-methylpropyl ketone

  • MDL number MFCD07699107
  • Cas Number : 898772-84-4
  • Molecular Weight : 240.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-12
Availability details for
Quantity
  • Availability
  • Add to Cart