RIE156321146 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl 1-methylbutyl ketone

  • MDL number MFCD07699110
  • Cas Number : 898772-90-2
  • Molecular Weight : 254.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-15
Availability details for
Quantity
  • Availability
  • Add to Cart