RIE156321150 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl 1-ethylpentyl ketone

  • MDL number MFCD07699114
  • Cas Number : 898772-98-0
  • Molecular Weight : 282.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-19
Availability details for
Quantity
  • Availability
  • Add to Cart