RIE156321151 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl 1-propylbutyl ketone

  • MDL number MFCD07699115
  • Cas Number : 898773-00-7
  • Molecular Weight : 282.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-20
Availability details for
Quantity
  • Availability
  • Add to Cart