RIE156321155 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(3-methoxybenzoyl)thiophene

  • MDL number MFCD07699119
  • Cas Number : 898773-11-0
  • Molecular Weight : 290.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [5-(1,3-dioxolan-2-yl)-2-thienyl](3-methoxyphenyl)methanone
Inchi Code 1S/C15H14O4S/c1-17-11-4-2-3-10(9-11)14(16)12-5-6-13(20-12)15-18-7-8-19-15/h2-6,9,15H,7-8H2,1H3
Inchi Key AJPSMODGWNQMMN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-03
Availability details for
Quantity
  • Availability
  • Add to Cart