RIE156321156 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(4-methoxybenzoyl)thiophene

  • MDL number MFCD07699120
  • Cas Number : 898773-14-3
  • Molecular Weight : 290.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [5-(1,3-dioxolan-2-yl)-2-thienyl](4-methoxyphenyl)methanone
Inchi Code 1S/C15H14O4S/c1-17-11-4-2-10(3-5-11)14(16)12-6-7-13(20-12)15-18-8-9-19-15/h2-7,15H,8-9H2,1H3
Inchi Key PJUHHMLLPRAKPC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart