RIE156321164 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(3-fluorobenzoyl)thiophene

  • MDL number MFCD07699128
  • Cas Number : 898773-38-1
  • Molecular Weight : 278.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-12
Availability details for
Quantity
  • Availability
  • Add to Cart