RIE156321167 Rieke Metals, Inc.

2-(3-Chlororobenzoyl)-5-(1,3-Dioxolan-2-yl)thiophene

  • MDL number MFCD07699131
  • Cas Number : 898773-47-2
  • Molecular Weight : 294.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-15
Availability details for
Quantity
  • Availability
  • Add to Cart