RIE156321168 Rieke Metals, Inc.

2-(2-Bromobenzoyl)-5-(1,3-Dioxolan-2-yl)thiophene

  • MDL number MFCD07699133
  • Cas Number : 898778-01-3
  • Molecular Weight : 339.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-17
Availability details for
Quantity
  • Availability
  • Add to Cart