RIE156321172 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(3-Iodobenzoyl)thiophene

  • MDL number MFCD07699137
  • Cas Number : 898778-13-7
  • Molecular Weight : 386.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-21
Availability details for
Quantity
  • Availability
  • Add to Cart