RIE156321174 Rieke Metals, Inc.

2-(3-Cyanobenzoyl)-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD07699139
  • Cas Number : 898778-19-3
  • Molecular Weight : 285.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-{[5-(1,3-dioxolan-2-yl)-2-thienyl]carbonyl}benzonitrile
Inchi Code 1S/C15H11NO3S/c16-9-10-2-1-3-11(8-10)14(17)12-4-5-13(20-12)15-18-6-7-19-15/h1-5,8,15H,6-7H2
Inchi Key KFFVHASQMJBHMR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-24

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-24
Availability details for
Quantity
  • Availability
  • Add to Cart