RIE156321177 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(4-phenoxybenzoyl)thiophene

  • MDL number MFCD07699142
  • Cas Number : 898778-28-4
  • Molecular Weight : 352.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-28
Availability details for
Quantity
  • Availability
  • Add to Cart