RIE156321180 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(4-isopropylbenzoyl)thiophene

  • MDL number MFCD07699145
  • Cas Number : 898778-37-5
  • Molecular Weight : 302.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [5-(1,3-dioxolan-2-yl)-2-thienyl](4-isopropylphenyl)methanone
Inchi Code 1S/C17H18O3S/c1-11(2)12-3-5-13(6-4-12)16(18)14-7-8-15(21-14)17-19-9-10-20-17/h3-8,11,17H,9-10H2,1-2H3
Inchi Key BUDNKTJZUJTFNW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-32
Availability details for
Quantity
  • Availability
  • Add to Cart