RIE156321182 Rieke Metals, Inc.

2-(4-t-Butylbenzoyl)-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD07699147
  • Cas Number : 898778-43-3
  • Molecular Weight : 316.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-34
Availability details for
Quantity
  • Availability
  • Add to Cart