RIE156321183 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(4-ethylbenzoyl)thiophene

  • MDL number MFCD07699148
  • Cas Number : 898778-46-6
  • Molecular Weight : 288.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-35A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-35A
Availability details for
Quantity
  • Availability
  • Add to Cart