RIE156321184 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(4-propylbenzoyl)thiophene

  • MDL number MFCD07699149
  • Cas Number : 898778-49-9
  • Molecular Weight : 302.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [5-(1,3-dioxolan-2-yl)-2-thienyl](4-propylphenyl)methanone
Inchi Code 1S/C17H18O3S/c1-2-3-12-4-6-13(7-5-12)16(18)14-8-9-15(21-14)17-19-10-11-20-17/h4-9,17H,2-3,10-11H2,1H3
Inchi Key OJIAKNAYNACWJZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-35b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-35b
Availability details for
Quantity
  • Availability
  • Add to Cart