RIE156321185 Rieke Metals, Inc.

2-(4-n-Butylbenzoyl)-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD07699150
  • Cas Number : 898778-52-4
  • Molecular Weight : 316.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-butylphenyl)[5-(1,3-dioxolan-2-yl)-2-thienyl]methanone
Inchi Code 1S/C18H20O3S/c1-2-3-4-13-5-7-14(8-6-13)17(19)15-9-10-16(22-15)18-20-11-12-21-18/h5-10,18H,2-4,11-12H2,1H3
Inchi Key YFNGYJXDWSOXJK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-35c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-35c
Availability details for
Quantity
  • Availability
  • Add to Cart