RIE156321187 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(4-hexylbenzoyl)thiophene

  • MDL number MFCD07699152
  • Cas Number : 898778-57-9
  • Molecular Weight : 344.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-35E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-35E
Availability details for
Quantity
  • Availability
  • Add to Cart