RIE156321190 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(4-propoxybenzoyl)thiophene

  • MDL number MFCD07699155
  • Cas Number : 898778-63-7
  • Molecular Weight : 318.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-36B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-36B
Availability details for
Quantity
  • Availability
  • Add to Cart