RIE156321191 Rieke Metals, Inc.

2-(4-n-Butoxybenzoyl)-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD07699156
  • Cas Number : 898778-65-9
  • Molecular Weight : 332.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-butoxyphenyl)[5-(1,3-dioxolan-2-yl)-2-thienyl]methanone
Inchi Code 1S/C18H20O4S/c1-2-3-10-20-14-6-4-13(5-7-14)17(19)15-8-9-16(23-15)18-21-11-12-22-18/h4-9,18H,2-3,10-12H2,1H3
Inchi Key UKCSOQRGMKYVOP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-36c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-36c
Availability details for
Quantity
  • Availability
  • Add to Cart