RIE156321193 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-(4-hexyloxybenzoyl)thiophene

  • MDL number MFCD07699158
  • Cas Number : 898778-69-3
  • Molecular Weight : 360.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [5-(1,3-dioxolan-2-yl)-2-thienyl][4-(hexyloxy)phenyl]methanone
Inchi Code 1S/C20H24O4S/c1-2-3-4-5-12-22-16-8-6-15(7-9-16)19(21)17-10-11-18(25-17)20-23-13-14-24-20/h6-11,20H,2-5,12-14H2,1H3
Inchi Key IHQPNVIECFFVGT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-36e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-36e
Availability details for
Quantity
  • Availability
  • Add to Cart