RIE156321201 Rieke Metals, Inc.

2-(2,3-Dichlorobenzoyl)-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD07699166
  • Cas Number : 898778-86-4
  • Molecular Weight : 329.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-43
Availability details for
Quantity
  • Availability
  • Add to Cart