RIE156321204 Rieke Metals, Inc.

2-(2,6-Dichlorobenzoyl)-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD07699169
  • Cas Number : 898778-92-2
  • Molecular Weight : 329.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,6-dichlorophenyl)[5-(1,3-dioxolan-2-yl)-2-thienyl]methanone
Inchi Code 1S/C14H10Cl2O3S/c15-8-2-1-3-9(16)12(8)13(17)10-4-5-11(20-10)14-18-6-7-19-14/h1-5,14H,6-7H2
Inchi Key JYCBJHNTXYUCOF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-46

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-46
Availability details for
Quantity
  • Availability
  • Add to Cart