RIE156321208 Rieke Metals, Inc.

2-(2,4-Dimethoxybenzoyl)-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD07699173
  • Cas Number : 898779-01-6
  • Molecular Weight : 320.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5568-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-50
Availability details for
Quantity
  • Availability
  • Add to Cart