RIE156321210 Rieke Metals, Inc.

2-(2,6-Dimethoxybenzoyl)-5-(1,3-dioxolan-2-yl)thiophene

  • MDL number MFCD07699175
  • Cas Number : 898779-07-2
  • Molecular Weight : 320.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,6-dimethoxyphenyl)[5-(1,3-dioxolan-2-yl)-2-thienyl]methanone
Inchi Code 1S/C16H16O5S/c1-18-10-4-3-5-11(19-2)14(10)15(17)12-6-7-13(22-12)16-20-8-9-21-16/h3-7,16H,8-9H2,1-2H3
Inchi Key RKGBBCPUJWQBRI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-52
Availability details for
Quantity
  • Availability
  • Add to Cart