RIE156321218 Rieke Metals, Inc.

3,4-Dichloro-4'-(ethylthio)benzophenone

  • MDL number MFCD06201550
  • Cas Number : 844885-36-5
  • Molecular Weight : 311.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5203-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5203-47
Availability details for
Quantity
  • Availability
  • Add to Cart