RIE156321219 Rieke Metals, Inc.

2-chloro-5-(4-ethoxycarbonylbenzoyl)pyridine

  • MDL number MFCD02260459
  • Cas Number : 727409-20-3
  • Molecular Weight : 289.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5514

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5514
Availability details for
Quantity
  • Availability
  • Add to Cart