RIE156321220 Rieke Metals, Inc.

2-chloro-5-(3-methyl-2-thenoyl)pyridine

  • MDL number MFCD02260464
  • Cas Number : 884504-82-9
  • Molecular Weight : 237.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5519

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5519
Availability details for
Quantity
  • Availability
  • Add to Cart