RIE156321222 Rieke Metals, Inc.

2-chloro-5-(2-thenoyl)pyridine

  • MDL number MFCD02260468
  • Cas Number : 501681-39-6
  • Molecular Weight : 223.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5523

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5523
Availability details for
Quantity
  • Availability
  • Add to Cart