RIE156321224 Rieke Metals, Inc.

4-Bromo-3',4'-dimethoxybenzophenone

  • MDL number MFCD02127292
  • Cas Number : 116412-90-9
  • Molecular Weight : 321.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5194-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5194-19
Availability details for
Quantity
  • Availability
  • Add to Cart