RIE156321226 Rieke Metals, Inc.

2-chloro-5-(5-ethoxycarbonyl-2-furoyl)pyridine

  • MDL number MFCD02260474
  • Cas Number : 884504-84-1
  • Molecular Weight : 279.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5529

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5529
Availability details for
Quantity
  • Availability
  • Add to Cart