RIE156321227 Rieke Metals, Inc.

4-Chloro-3'-fluorobenzophenone

  • MDL number MFCD09801597
  • Cas Number : 46698-36-6
  • Molecular Weight : 234.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5204-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5204-16
Availability details for
Quantity
  • Availability
  • Add to Cart