RIE156321231 Rieke Metals, Inc.

Ethyl 4-(2-chloro-3-pyridyl)-4-oxobutyrate

  • MDL number MFCD02260480
  • Cas Number : 852063-32-2
  • Molecular Weight : 241.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5535B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5535B
Availability details for
Quantity
  • Availability
  • Add to Cart