RIE156321233 Rieke Metals, Inc.

3,4-Dimethoxy-4'-iodobenzophenone

  • MDL number MFCD00475359
  • Cas Number : 116413-00-4
  • Molecular Weight : 368.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5194-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5194-22
Availability details for
Quantity
  • Availability
  • Add to Cart