RIE156321236 Rieke Metals, Inc.

3,4-Difluoro-3',4'-dimethoxybenzophenone

  • MDL number MFCD06201564
  • Cas Number : 845781-07-9
  • Molecular Weight : 278.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5194-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5194-41
Availability details for
Quantity
  • Availability
  • Add to Cart