RIE156321238 Rieke Metals, Inc.

3-Chloro-4'-fluorobenzophenone

  • MDL number MFCD09801606
  • Cas Number : 2192-35-0
  • Molecular Weight : 234.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5205-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5205-15
Availability details for
Quantity
  • Availability
  • Add to Cart