RIE156321248 Rieke Metals, Inc.

4-tert-Butyl-3',5'-difluorobenzophenone

  • MDL number MFCD06201517
  • Cas Number : 844885-10-5
  • Molecular Weight : 274.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5181-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5181-42
Availability details for
Quantity
  • Availability
  • Add to Cart