RIE156321249 Rieke Metals, Inc.

3-Bromo-3'-fluoro-4'-methoxybenzophenone

  • MDL number MFCD06201474
  • Cas Number : 844879-54-5
  • Molecular Weight : 309.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)(3-fluoro-4-methoxyphenyl)methanone
Inchi Code 1S/C14H10BrFO2/c1-18-13-6-5-10(8-12(13)16)14(17)9-3-2-4-11(15)7-9/h2-8H,1H3
Inchi Key XFBBZIQLOGYHLY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5206-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5206-18
Availability details for
Quantity
  • Availability
  • Add to Cart