RIE156321258 Rieke Metals, Inc.

2,4'-Dichlorobenzophenone

  • MDL number MFCD00038744
  • Cas Number : 85-29-0
  • Molecular Weight : 251.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5182-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5182-16
Availability details for
Quantity
  • Availability
  • Add to Cart