RIE156321260 Rieke Metals, Inc.

3-chloro-3'-nitrobenzophenone

  • MDL number MFCD00228117
  • Cas Number : 494203-12-2
  • Molecular Weight : 261.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5087

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5087
Availability details for
Quantity
  • Availability
  • Add to Cart