RIE156321264 Rieke Metals, Inc.

3-(2-Thenoyl)pyridine

  • MDL number MFCD03841023
  • Cas Number : 21327-72-0
  • Molecular Weight : 189.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-pyridinyl(2-thienyl)methanone
Inchi Code 1S/C10H7NOS/c12-10(9-4-2-6-13-9)8-3-1-5-11-7-8/h1-7H
Inchi Key OQWPFCKVCCDORA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5552

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5552
Availability details for
Quantity
  • Availability
  • Add to Cart